Industrial gloves, Edge® 48-126
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
48-126/11 48-126/9 48-126/6 48-126/8 48-126/7 48-126/10
ANSE48-126/11EA 188 DKK
ANSE48-126/11 ANSE48-126/9 ANSE48-126/6 ANSE48-126/8 ANSE48-126/7 ANSE48-126/10
Industrial gloves, Edge® 48-126
Handsker
These knitted polyester liners with PU dip coating provide a good level of abrasion resistance and grip, as well as high tactility in dry to slightly oily environments.
  • 15 gauge highly comfortable and dexterous polyester liner
  • Seamless knitted liner with elasticized knit wrist dipped into white polyurethane coating
  • Good flexibility and finger dexterity compared to cotton canvas or thick cotton gloves
  • Dark colour shows dirt less, glove is discarded less quickly
Recommended for: Assembly of small pieces, general handling, packaging, quality inspection.
Certificeringer: EN 388:2016 (4121A); Cat. II
Emballering: 12 pairs per bag.