meta-Phosphorsyre, lumps, AnalaR NORMAPUR® analytical reagent

Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Metaphosphoric acid

20632.236EA 817 DKK
20632.236 20632.293
meta-Phosphorsyre, lumps, AnalaR NORMAPUR® analytical reagent
meta-Phosphorsyre

Mixture of metaphosphoric acid and sodium metaphosphate (NaPO3 : 56 to 60%)

Formel: (HPO₃)n
Molekylvægt: 79.98 g/mol
Kogepunkt: 260
Smeltepunkt: 21
Massefylde: 2.4 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011351
CAS-nummer: 37267-86-0
EINECS: 253-433-4
UN nummer: 3260
ADR: 8 III

Bestil nu

Specifikationer Testresultater

Assay (calculated as HPO3) (39-45 %) Passes test
Assay (calculated as NaPO3) (55-61 %) Passes test
Heavy metals (as Pb) Max. 20 ppm
Substances reducing KMnO4 (as H3PO3) Max. 100 ppm
Cl (Chloride) Max. 20 ppm
SO4 (Sulphate) Max. 100 ppm
As (Arsenic) Max. 5 ppm
Ca (Calcium) Max. 100 ppm
Fe (Iron) Max. 10 ppm
Mn (Manganese) Max. 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR