ω-Brom-4-methylacetophenon 97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
170390050
170390250
170391000
ACRO170390050EA
358
DKK
ACRO170390050
ACRO170390250
ACRO170391000
ω-Brom-4-methylacetophenon 97%
ω-Brom-4-methylacetophenon
Formel:
C₉H₉BrO Kogepunkt: 105 °C (0,1 -rr) Smeltepunkt: 52…53 °C Flammepunkt: >110 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00000203 CAS-nummer: 619-41-0 EINECS: 210-595-0 |
Specifikationer Testresultater
Appearance | White to light yellow Crystalline powder or needles |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...