α,α-Dimethoxytoluen 98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Benzaldehyde dimethyl acetal
209791000
209792500
209790010
209790025
ACRO209791000EA
335
DKK
ACRO209791000
ACRO209792500
ACRO209790010
ACRO209790025
α,α-Dimethoxytoluen 98%
α,α-Dimethoxytoluen
Formel:
C₉H₁₂O₂ Kogepunkt: 87…89 °C (18 mmHg) Massefylde: 1,01 g/cm³ Flammepunkt: 69 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00008491 CAS-nummer: 1125-88-8 EINECS: 214-413-0 |
Specifikationer Testresultater
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4910 to 1.4940 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...