3-iod-p-toluidin ≥98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
A11485.06
A11485.14
A11485.22
A11485.06EA
456
DKK
A11485.06
A11485.14
A11485.22
3-iod-p-toluidin ≥98%
3-iod-p-toluidin
Purity: 98%
CAS No.: 35944-64-0
Molecular Formula: C7H8IN
Formula Weight: 233.05
Melting Point: 34-37°
Beilstein Reference: 2353567
UN No.: 2811
MDL No.: MFCD00047843
Formel:
C₇H₈IN Molekylvægt: 233.05 g/mol Smeltepunkt: 34…37 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00047843 CAS-nummer: 35944-64-0 EINECS: 252-803-2 UN nummer: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...