2'-Fluor-4'-hydroxyacetophenon ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-(2-Fluoro-4-hydroxyphenyl)ethanone

A19884.06 A19884.14
A19884.06EA 1210 DKK
A19884.06 A19884.14
2'-Fluor-4'-hydroxyacetophenon ≥97%
2'-Fluor-4'-hydroxyacetophenon

Purity: 97% CAS No.: 98619-07-9 Molecular Formula: C8H7FO2 Formula Weight: 154.14 Melting Point: 121-123° Beilstein Reference: 696642

Formel: C₈H₇FO₂
Molekylvægt: 154.14 g/mol
Smeltepunkt: 121…123 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00142713
CAS-nummer: 98619-07-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR