4-(Chlormethyl)benzoylchlorid ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
C1283-25G
C1283-500G
C1283-100G
TCIAC1283-25GEA
181
DKK
TCIAC1283-25G
TCIAC1283-500G
TCIAC1283-100G
4-(Chlormethyl)benzoylchlorid ≥98.0% (by GC, titration analysis)
4-(Chlormethyl)benzoylchlorid
Formel:
C₈H₆Cl₂O Kogepunkt: 276 °C (1013 hPa) Smeltepunkt: 30…31 °C Massefylde: 1,3034 g/cm³ (20 °C) |
MDL Number:
MFCD00053224 CAS-nummer: 876-08-4 EINECS: 212-881-0 UN nummer: 3261 ADR: 8,II |
|
Specifikationer Testresultater
Appearance | White or Colorless to Almost white or Almost colorless powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Solubility in Toluene | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...