3'-Brom-4'-Fluoracetophenon ≥98%
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
B22091.14 B22091.06
B22091.14EA 3140 DKK
B22091.14 B22091.06
3'-Brom-4'-Fluoracetophenon ≥98%
Purity: 98+% CAS No.: 1007-15-4 Molecular Formula: C8H6BrFO Formula Weight: 217.04 Flash Point: 110°(230°F) Melting Point: 55-57° Boiling Point: 150°/12mm Beilstein Reference: 3088698 MDL No.: MFCD00042466
Formel: C₈H₆BrFO
Kogepunkt: 150 °C (12 mmHg)
Smeltepunkt: 55…57 °C
Flammepunkt: >110 °C (230 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00042466
CAS-nummer: 1007-15-4
EINECS: 213-750-0