2'-Brom-4'-Fluoracetophenon ≥98%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Purity: 98%
CAS No.: 1006-39-9
Molecular Formula: C8H6BrFO
Formula Weight: 217.04
Flash Point:
Formel:
C₈H₆BrFO Kogepunkt: 150 °C (12 mmHg) Flammepunkt: >110 °C (230 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00077464 CAS-nummer: 1006-39-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...