Benzensulfonamid ≥98%
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
A10891.0B A10891.30
A10891.0BEA 1100 DKK
A10891.0B A10891.30
Benzensulfonamid ≥98%
Benzensulfonamid
Purity: 98+% CAS No.: 98-10-2 Molecular Formula: C6H7NO2S Formula Weight: 157.19 Flash Point: 250°(482°F) Odour: Odourless Beilstein Reference: 1100566 MDL No.: MFCD00007930
Formel: C₆H₇NO₂S
Molekylvægt: 157.19 g/mol
Smeltepunkt: 151…154 °C
Flammepunkt: 250 °C (482 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007930
CAS-nummer: 98-10-2
EINECS: 202-637-1