5,5'-Dithiobis(2-nitrobenzosyre)

Leverandør: Serva
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: DTNB, Ellman's reagent, 3,3'-Dithiobis(6-nitrobenzoic acid), 5,5'-Disulphanediylbis(2-nitrobenzoic acid), 2,2'-Dinitro-5,5'-dithiodibenzoic acid

20735.02 20735.03
SERA20735.02EA 522.1 DKK
SERA20735.02 SERA20735.03
5,5'-Dithiobis(2-nitrobenzosyre)
5,5'-Dithiobis(2-nitrobenzosyre)
Formel: C₁₄H₈N₂O₈S₂
Molekylvægt: 396.36 g/mol
Smeltepunkt: 240…244 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007140
CAS-nummer: 69-78-3
EINECS: 200-714-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR