5,5'-Dithiobis(2-nitrobenzosyre) ≥99%
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: DTNB, Ellman's reagent, 3,3'-Dithiobis(6-nitrobenzoic acid), 5,5'-Disulphanediylbis(2-nitrobenzoic acid), 2,2'-Dinitro-5,5'-dithiodibenzoic acid
A14331.03 A14331.06 A14331.14
A14331.03EA 343 DKK
A14331.03 A14331.06 A14331.14
5,5'-Dithiobis(2-nitrobenzosyre) ≥99%
5,5'-Dithiobis(2-nitrobenzosyre)
Purity: 99% CAS No.: 69-78-3 Molecular Formula: C14H8N2O8S2 Formula Weight: 396.36 Melting Point: ca 240° dec. Beilstein Reference: 1896821
Formel: C₁₄H₈N₂O₈S₂
Molekylvægt: 396.36 g/mol
Smeltepunkt: ∼ 240 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007140
CAS-nummer: 69-78-3
EINECS: 200-714-4