Dimethyl 5-hydroxyisophthalat ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 5-Hydroxyisophthalic acid dimethyl ester, Dimethyl 5-hydroxyisophthalate

B21565.18 B21565.09 B21565.30
B21565.18EA 1310 DKK
B21565.18 B21565.09 B21565.30
Dimethyl 5-hydroxyisophthalat ≥98%
Dimethyl 5-hydroxyisophthalat

Purity: 98% CAS No.: 13036-02-7 Molecular Formula: C10H10O5 Formula Weight: 210.19 Melting Point: 162-164° Beilstein Reference: 2650120

Formel: C₁₀H₁₀O₅
Smeltepunkt: 162…164 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00134367
CAS-nummer: 13036-02-7
EINECS: 235-899-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR