Dimethyl 5-hydroxyisophthalat 99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
5-Hydroxyisophthalic acid dimethyl ester, Dimethyl 5-hydroxyisophthalate
356440100
356440250
ACRO356440100EA
370
DKK
ACRO356440100
ACRO356440250
Dimethyl 5-hydroxyisophthalat 99%
Dimethyl 5-hydroxyisophthalat
Formel:
C₁₀H₁₀O₅ Smeltepunkt: 162…165 °C |
MDL Number:
MFCD00134367 CAS-nummer: 13036-02-7 EINECS: 235-899-0 |
Specifikationer Testresultater
Appearance | White to light beige Powder and/or lumps |
Infrared spectrum | Conforms |
Melting point | 162 to 165 °C |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...