Zinc 2-Mercaptobenzothiazole ≥97.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Mercaptobenzothiazolezincsalt, 2-Mercaptobenzothiazole Zinc Salt, Zinc 2-Benzothiazolethiolate

Z0033-25G Z0033-500G
TCIAZ0033-25GEA 132 DKK
TCIAZ0033-25G TCIAZ0033-500G
Zinc 2-Mercaptobenzothiazole ≥97.0% (by HPLC, titration analysis)
Zinc 2-Mercaptobenzothiazole
Formel: C₁₄H₈N₂S₄Zn
MDL Number: MFCD00072234
CAS-nummer: 155-04-4
EINECS: 205-840-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45b63901871f7f51eda5b0c39f1572eb3831c5c3&vid=45b63901871f7f51eda5b0c39f1572eb3831c5c3&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Chelometric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR