Zonisamide ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1,2-Benzisoxazole-3-methanesulphonamide, Benzo[d]isoxazol-3-ylmethanesulphonamide

Z0026-200MG Z0026-1G
TCIAZ0026-200MGEA 271 DKK
TCIAZ0026-200MG TCIAZ0026-1G
Zonisamide ≥98.0% (by HPLC, total nitrogen)
Zonisamide
Formel: C₈H₈N₂O₃S
Molekylvægt: 212.22 g/mol
MDL Number: MFCD00865316
CAS-nummer: 68291-97-4
Merck Index: 14,10192
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=051c83777afa5b1791b27dea716c4bdc73f1ced5&vid=051c83777afa5b1791b27dea716c4bdc73f1ced5&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 163.0 to 167.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR