Zebularine ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Z0022-1G Z0022-200MG
TCIAZ0022-1GEA 1510 DKK
TCIAZ0022-1G TCIAZ0022-200MG
Zebularine ≥98.0% (by HPLC, titration analysis)
Zebularine
Formel: C₉H₁₂N₂O₅
Opbevaringstemperatur: Køleskab
MDL Number: MFCD04973699
CAS-nummer: 3690-10-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b4427bbc1813ab54e5854619cb2ec740a6ecda1f&vid=b4427bbc1813ab54e5854619cb2ec740a6ecda1f&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Specific rotation [a]20/D +156 ~ +166 deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR