Zirconocene bis(trifluoromethanesulphonate) tetrahydrofuran adduct ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Bis(cyclopentadienyl)zirconium Bis(trifluoromethanesulphonate), Zirconocene Bis(triflate)

Z0009-1G Z0009-5G
TCIAZ0009-1GEA 1010 DKK
TCIAZ0009-1G TCIAZ0009-5G
Zirconocene bis(trifluoromethanesulphonate) tetrahydrofuran adduct ≥97.0% (by titrimetric analysis)
Zirconocene bis(trifluoromethanesulphonate) tetrahydrofuran
Formel: C₁₆H₁₈F₆O₇S₂Zr
Molekylvægt: 591.65 g/mol
MDL Number: MFCD00192583
CAS-nummer: 89672-77-5
UN nummer: 1325
ADR: 4.1 III

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (Chelometric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR