Zein (from corn)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Z0001-25G Z0001-100G
TCIAZ0001-25GEA 189 DKK
TCIAZ0001-25G TCIAZ0001-100G
Zein (from corn)
Zein
MDL Number: MFCD00062414
CAS-nummer: 9010-66-6
EINECS: 232-722-9
Merck Index: 14,10119

Bestil nu

Specifikationer Testresultater

Appearance White to Yellow to Orange powder to crystal
Total Nitrogen 13.0 to 16.0 %
Drying loss max. 5.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR