Ytterbium(III) isopropoxide ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Ytterbium(III) i-propoxide

Y0010-200MG Y0010-1G
TCIAY0010-200MGEA 419 DKK
TCIAY0010-200MG TCIAY0010-1G
Ytterbium(III) isopropoxide ≥98.0% (by titrimetric analysis)
Ytterbium(III) isopropoxide
Formel: C₉H₂₁O₃Yb
Molekylvægt: 350.30 g/mol
MDL Number: MFCD00145532
CAS-nummer: 6742-69-4
UN nummer: 2925
ADR: 4.1 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e72408616bc336c17a43241920f02793104fadf3&vid=1df27f6aa783a310a2e9b7f8b5b1c5ae53aa7821&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR