2-Vinylphenylboronsyre (contains varying amounts of Anhydride)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Dihydroxy(2-vinylphenyl)borane, (2-Vinylphenyl)metaboric acid, 2-Vinylbenzeneboronic acid
V0140-1G
V0140-5G
TCIAV0140-1GEA
665
DKK
TCIAV0140-1G
TCIAV0140-5G
2-Vinylphenylboronsyre (contains varying amounts of Anhydride)
2-Vinylphenylboronsyre
Formel:
C₈H₉BO₂ Molekylvægt: 147.97 g/mol Smeltepunkt: 109°C |
MDL Number:
MFCD01074659 CAS-nummer: 15016-42-9 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 114.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...