4-Vinylbenzylamine ≥92.0% (by GC) stabilised

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (4-Vinylphenyl)methanamine, 4-(Aminomethyl)styrene

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

V0101-1G V0101-5G
TCIAV0101-1GEA 870 DKK
TCIAV0101-1G TCIAV0101-5G
4-Vinylbenzylamine ≥92.0% (by GC) stabilised
4-Vinylbenzylamine
Formel: C₉H₁₁N
Molekylvægt: 133.19 g/mol
Kogepunkt: 60°C
Opbevaringstemperatur: Fryser
MDL Number: MFCD08704374
CAS-nummer: 50325-49-0

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 92.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR