D-Valine benzyl ester-p-toluenesulphonate ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
H-D-Val-OBzl.Tos-OH, D-Valine Benzyl Ester p-toluenesulphonate, H-D-Val-Obzl.TosOH
V0095-1G
V0095-5G
TCIAV0095-1GEA
222
DKK
TCIAV0095-1G
TCIAV0095-5G
D-Valine benzyl ester-p-toluenesulphonate ≥98.0% (by HPLC, titration analysis)
D-Valine benzyl ester-p-toluenesulphonate
Formel:
C₁₂H₁₇NO₂ Molekylvægt: 379.47 g/mol |
MDL Number:
MFCD00136545 CAS-nummer: 17662-84-9 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 160.0 to 164.0 deg-C |
Specific rotation [a]20/D | +2.0 to +5.0 deg (C=3.2, methanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...