N-Vinylphthalimide ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
V0065-25G
V0065-5G
TCIAV0065-25GEA
2660
DKK
TCIAV0065-25G
TCIAV0065-5G
N-Vinylphthalimide ≥98.0% (by GC)
N-Vinylphthalimide
Formel:
C₁₀H₇NO₂ Kogepunkt: 151°C |
MDL Number:
MFCD00078446 CAS-nummer: 3485-84-5 EINECS: 222-473-4 |
Specifikationer Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 82.0 to 87.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...