L-Valine methyl ester hydrochloride ≥98.5% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: H-Val-OMe.HCl, L-2-Aminoisovaleric Acid Methyl Ester Hydrochloride, L-Methyl valinate hydrochloride, H-Val-OMe·HCl

V0056-25G V0056-5G
TCIAV0056-25GEA 435 DKK
TCIAV0056-25G TCIAV0056-5G
L-Valine methyl ester hydrochloride ≥98.5% (by titrimetric analysis)
L-Valine methyl ester hydrochloride
Formel: C₆H₁₄ClNO₂
Molekylvægt: 167.63 g/mol
MDL Number: MFCD00012497
CAS-nummer: 6306-52-1
EINECS: 228-620-9

Bestil nu

Specifikationer Testresultater

Appearance White powder to crystal
Purity (Argentmetric Titration) min. 98.5 %
Specific rotation [a]20/D +15.0 to +16.0 deg (C=2, H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR