Ursolic acid ≥90.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 3beta-Hydroxy-12-ursen-28-oic Acid

U0065-1G U0065-100MG
TCIAU0065-1GEA 1990 DKK
TCIAU0065-1G TCIAU0065-100MG
Ursolic acid ≥90.0% (by GC)
Ursolic acid
Formel: C₃₀H₄₈O₃
MDL Number: MFCD00009621
CAS-nummer: 77-52-1
EINECS: 201-034-0
Merck Index: 14,09890

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 90.0 %
Specific rotation [a]20/D +58.0 to +65.0 deg (C=1, Pyridine)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR