1,3,5-Undecatriene (mixture of isomers) ≥75.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
U0062-5ML
U0062-25ML
TCIAU0062-5MLEA
517
DKK
TCIAU0062-5ML
TCIAU0062-25ML
1,3,5-Undecatriene (mixture of isomers) ≥75.0% (by GC)
1,3,5-Undecatriene (mixture of isomers)
Formel:
C₁₁H₁₈ Molekylvægt: 150.27 g/mol Flammepunkt: 65°C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00015699 CAS-nummer: 16356-11-9 EINECS: 240-416-1 |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 75.0% (total of isomer) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...