Trimethyl(4-vinylphenyl)silane ≥95.0% (by GC) stabilised
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
1-Ethenyl-4-(trimethylsilyl)benzene, 4-Trimethylsilylstyrene
Stabilization:
Stabilised with 4-tert-butylpyrocatechol (TBC)
T3919-1G
T3919-250MG
TCIAT3919-1GEA
920
DKK
TCIAT3919-1G
TCIAT3919-250MG
Trimethyl(4-vinylphenyl)silane ≥95.0% (by GC) stabilised
Trimethyl(4-vinylphenyl)silane
Formel:
C₁₁H₁₆Si Molekylvægt: 176.33 g/mol Kogepunkt: 90°C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD05663945 CAS-nummer: 1009-43-4 |
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 95.0 % |
Specific gravity (20/20) | 0.8880 to 0.9020 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...