Tetrakisacetonitrile copper(I) triflate ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Tetrakis(acetonitrile)copper trifluoromethanesulphonate, Tetrakis(acetonitrile)copper(I) Trifluoromethanesulphonate
T3905-1G
T3905-5G
TCIAT3905-1GEA
534
DKK
TCIAT3905-1G
TCIAT3905-5G
Tetrakisacetonitrile copper(I) triflate ≥98.0% (by titrimetric analysis)
Tetrakisacetonitrile copper(I) triflate
Formel:
Cu(NCCH₃)₄·CF₃SO₃ Smeltepunkt: 124°C |
MDL Number:
MFCD10566992 CAS-nummer: 58452-28-1 |
Specifikationer Testresultater
Appearance | White to Light yellow to Green powder to crystal |
Purity (Potassium permanganate method) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...