N-(1,1,3,3-Tetramethylbutyl)acrylamide ≥95.0% (by GC) stabilised

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

T3861-25G T3861-100G
TCIAT3861-25GEA 591 DKK
TCIAT3861-25G TCIAT3861-100G
N-(1,1,3,3-Tetramethylbutyl)acrylamide ≥95.0% (by GC) stabilised
N-(1,1,3,3-Tetramethylbutyl)acrylamide
Formel: C₁₁H₂₁NO
Kogepunkt: 142°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00048259
CAS-nummer: 4223-03-4
EINECS: 224-169-7

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to lump
Purity (GC) min. 95.0 %
Melting point 61.0 to 65.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR