Triethoxy(p-tolyl)silane ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

T3750-1G T3750-5G
TCIAT3750-1GEA 296 DKK
TCIAT3750-1G TCIAT3750-5G
Triethoxy(p-tolyl)silane ≥95.0% (by GC)
Triethoxy(p-tolyl)silane
Formel: C₁₃H₂₂O₃Si
Molekylvægt: 254.40 g/mol
Kogepunkt: 106°C
MDL Number: MFCD04974099
CAS-nummer: 18412-57-2

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR