3-(Trifluoromethyl)cyclohexylamine cis- and trans-mixture ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 3-(Trifluoromethyl)cyclohexanamine

T3678-1G T3678-5G
TCIAT3678-1GEA 838 DKK
TCIAT3678-1G TCIAT3678-5G
3-(Trifluoromethyl)cyclohexylamine cis- and trans-mixture ≥98.0% (by GC, titration analysis)
3-(Trifluoromethyl)cyclohexylamine
Formel: C₇H₁₂F₃N
Kogepunkt: 70°C
MDL Number: MFCD08444665
CAS-nummer: 56287-83-3
UN nummer: 2734
ADR: 8,II

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 % (total of isomer)
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR