Tetrabutylammoniumnitrat ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

T3651-25G T3651-5G
TCIAT3651-25GEA 649 DKK
TCIAT3651-25G TCIAT3651-5G
Tetrabutylammoniumnitrat ≥98.0% (by titrimetric analysis)
Tetrabutylammonium nitrate
Formel: C₁₆H₃₆N₂O₃
Molekylvægt: 304.47 g/mol
MDL Number: MFCD00043202
CAS-nummer: 1941-27-1
EINECS: 217-726-0
UN nummer: 1479
ADR: 5.1 II

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 117.0 to 121.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR