(Heptafluoropropyl)trimethylsilane ≥98.0% (by GC)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: Trimethyl(heptafluoropropyl)silane, Trimethyl(perfluoropropyl)silane
T3593-1G T3593-5G
TCIAT3593-1GEA 805 DKK
TCIAT3593-1G TCIAT3593-5G
(Heptafluoropropyl)trimethylsilane ≥98.0% (by GC)
(Heptafluoropropyl)trimethylsilane
Formel: C₆H₉F₇Si
Kogepunkt: 88°C
Flammepunkt: 54°C
MDL Number: MFCD00216712
CAS-nummer: 3834-42-2
UN nummer: 1993
ADR: 3,III
Bestil nu
Specifikationer Testresultater
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %