Trimethylsilyl acrylate ≥95.0% (by GC) stabilised
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Acrylic acid, Trimethylsilyl Acrylate, Acrylic acid trimethylsilyl ester, Acryloxytrimethylsilane
Stabilization:
Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
T3559-5ML
T3559-25ML
TCIAT3559-5MLEA
312
DKK
TCIAT3559-5ML
TCIAT3559-25ML
Trimethylsilyl acrylate ≥95.0% (by GC) stabilised
Trimethylsilyl acrylate
Formel:
C₆H₁₂O₂Si Molekylvægt: 144.25 g/mol Kogepunkt: 54°C Flammepunkt: –15°C |
MDL Number:
MFCD00156432 CAS-nummer: 13688-55-6 UN nummer: 1993 ADR: 3 II |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...