Trimethylsilyl Propionate ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Propionic acid trimethylsilyl ester

T3552-1ML T3552-5ML
TCIAT3552-1MLEA 386 DKK
TCIAT3552-1ML TCIAT3552-5ML
Trimethylsilyl Propionate ≥95.0% (by GC)
Trimethylsilyl Propionate
Formel: C₆H₁₄O₂Si
Kogepunkt: 122°C
MDL Number: MFCD01632555
CAS-nummer: 16844-98-7
UN nummer: 1993
ADR: 3,II

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR