2,4,6-Tri([1,1'-biphenyl]-4-yl)-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
T3539-200MG
T3539-1G
TCIAT3539-200MGEA
567
DKK
TCIAT3539-200MG
TCIAT3539-1G
2,4,6-Tri([1,1'-biphenyl]-4-yl)-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)
2,4,6-Tri([1,1'-biphenyl]-4-yl)-1,3,5-triazine
Formel:
C₃₉H₂₇N₃ Molekylvægt: 537.67 g/mol |
CAS-nummer:
31274-51-8 |
|
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...