3-(2-Thienyl)-L-alanine ≥97.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: H-β-(2-Thienyl)-Ala-OH, 3-(2-Thienyl)-L-alanine, (S)-2-Amino-3-(thiophen-2-yl)propanoic acid

T3475-200MG T3475-1G
TCIAT3475-200MGEA 279 DKK
TCIAT3475-200MG TCIAT3475-1G
3-(2-Thienyl)-L-alanine ≥97.0% (by HPLC, titration analysis)
3-(2-Thienyl)-L-alanine
Formel: C₇H₉NO₂S
Molekylvægt: 171.214 g/mol
MDL Number: MFCD00064335
CAS-nummer: 22951-96-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=61ba3c6353ad3a51dc173a21f77c305337775fa1&vid=61ba3c6353ad3a51dc173a21f77c305337775fa1&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %
Specific rotation [a]20/D -29.0 to -33.0 deg (C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR