Triethyleneglycol divinyl ether ≥98.0% (by GC) stabilised
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: 3,6,9,12-Tetraoxatetradeca-1,13-diene
Stabilization: Stabilised with potassium hydroxide
T3460-25G T3460-100G
TCIAT3460-25GEA 403 DKK
TCIAT3460-25G TCIAT3460-100G
Triethyleneglycol divinyl ether ≥98.0% (by GC) stabilised
Triethyleneglycol divinyl ether
Formel: C₁₀H₁₈O₄
Molekylvægt: 202.2475 g/mol
Kogepunkt: 126°C
MDL Number: MFCD00074891
CAS-nummer: 765-12-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=298236bfe44b9a6db79c61bb3db7665b30ae08cf&vid=298236bfe44b9a6db79c61bb3db7665b30ae08cf&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Specific gravity (20/20) 1.0020 to 1.0060
Refractive index n20/D 1.4510 to 1.4550