1,1,2,3-Tetramethyl-1H-benzo[e]indolium Hexafluorophosphate ≥97.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

T3457-5G T3457-25G
TCIAT3457-5GEA 427 DKK
TCIAT3457-5G TCIAT3457-25G
1,1,2,3-Tetramethyl-1H-benzo[e]indolium Hexafluorophosphate ≥97.0% (by HPLC, total nitrogen)
1,1,2,3-Tetramethyl-1H-benzo[e]indolium Hexafluorophosphate
Formel: C₁₆H₁₈F₆NP
MDL Number: MFCD16621101
CAS-nummer: 372081-65-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dbfe7ce7d79694ee0c7dde2ea4d70215c9aa5cae&vid=dbfe7ce7d79694ee0c7dde2ea4d70215c9aa5cae&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Purple to Brown powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR