4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzylamine hydrochloride ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3-[4-(Aminomethyl)phenyl]-3-(trifluoromethyl)-3H-diazirine hydrochloride

T3448-200MG T3448-1G
TCIAT3448-200MGEA 969 DKK
TCIAT3448-200MG TCIAT3448-1G
4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzylamine hydrochloride ≥98.0% (by HPLC, titration analysis)
4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzylamine hydrochloride
Formel: C₉H₈F₃N₃·HCl
Opbevaringstemperatur: Køleskab
MDL Number: MFCD29765752
CAS-nummer: 1258874-29-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d12e27fee3372182ca24571ea4a94ce3187fef19&vid=d12e27fee3372182ca24571ea4a94ce3187fef19&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR