1,3,5-Tris[4-[bis(4-methoxyphenyl)amino]phenyl]benzene ≥95.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

T3437-200MG T3437-1G
TCIAT3437-200MGEA 985 DKK
TCIAT3437-200MG TCIAT3437-1G
1,3,5-Tris[4-[bis(4-methoxyphenyl)amino]phenyl]benzene ≥95.0% (by HPLC)
1,3,5-Tris[4-[bis(4-methoxyphenyl)amino]phenyl]benzene
Formel: C₆₆H₅₇N₃O₆
Molekylvægt: 988.20 g/mol
CAS-nummer: 142894-38-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15556aa17edc8df0ca8258d1b7d6711dc0c14d28&vid=bd913949969520b89285d655327fc15603943197&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Elemental analysis (Nitrogen) 3.80 and le 4.70 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR