trans-3,4,5-Trifluoro-4'-(4-n-pentylcyclohexyl)biphenyl ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
3,4,5-Trifluoro-4'-(trans-4-pentylcyclohexyl)-1,1'-biphenyl, 3,4,5-Trifluoro-4'-(trans-4-pentylcyclohexyl)biphenyl
T3397-1G
T3397-5G
TCIAT3397-1GEA
444
DKK
TCIAT3397-1G
TCIAT3397-5G
trans-3,4,5-Trifluoro-4'-(4-n-pentylcyclohexyl)biphenyl ≥98.0% (by GC)
trans-3,4,5-Trifluoro-4'-(4-n-pentylcyclohexyl)biphenyl
Formel:
C₂₃H₂₇F₃ Molekylvægt: 360.464 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD09839002 CAS-nummer: 137019-95-5 |
Specifikationer Testresultater
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...