2,4,6-Triphenylpyridine ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
T3394-200MG
T3394-1G
TCIAT3394-200MGEA
403
DKK
TCIAT3394-200MG
TCIAT3394-1G
2,4,6-Triphenylpyridine ≥98.0% (by GC)
2,4,6-Triphenylpyridine
Formel:
C₂₃H₁₇N Molekylvægt: 307.40 g/mol |
MDL Number:
MFCD00014630 CAS-nummer: 580-35-8 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 139.0 to 143.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...