2-(Trifluormethyl)benzamid ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

T3384-5G T3384-25G
TCIAT3384-5GEA 542 DKK
TCIAT3384-5G TCIAT3384-25G
2-(Trifluormethyl)benzamid ≥98.0% (by GC)
2-(Trifluormethyl)benzamid
Formel: C₈H₆F₃NO
Molekylvægt: 189.14 g/mol
MDL Number: MFCD00014798
CAS-nummer: 360-64-5
EINECS: 206-637-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b05049cb9f073c59ca951336deeca737f5afe6b&vid=9b05049cb9f073c59ca951336deeca737f5afe6b&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 162.0 to 166.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR