Tetrabutylammonium tetrakis(trifluoromethyl)cuprate(III) ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Tetrabutylammonium tetrakis(trifluoromethyl)copper(III)

T3374-200MG T3374-1G
TCIAT3374-200MGEA 821 DKK
TCIAT3374-200MG TCIAT3374-1G
Tetrabutylammonium tetrakis(trifluoromethyl)cuprate(III) ≥98.0% (by titrimetric analysis)
Tetrabutylammonium tetrakis(trifluoromethyl)cuprate(III)
Formel: C₂₀H₃₆CuF₁₂N
Molekylvægt: 582.04 g/mol
Smeltepunkt: 142°C
CAS-nummer: 152376-94-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9de839079bccfaa8cca860ab2ea87b6eb4c18a4c&vid=9de839079bccfaa8cca860ab2ea87b6eb4c18a4c&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR