3-(Perfluoro-n-hexyl)-1,2-propenoxide ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2,2,3,3,4,4,5,5,6,6,7,7,7-Tridecafluoroheptyloxirane, 3-Perfluorohexyl-1,2-epoxypropane
T3365-5G
T3365-25G
TCIAT3365-5GEA
1290
DKK
TCIAT3365-5G
TCIAT3365-25G
3-(Perfluoro-n-hexyl)-1,2-propenoxide ≥98.0% (by GC)
3-(Perfluoro-n-hexyl)-1,2-propenoxide
Formel:
C₉H₅F₁₃O Kogepunkt: 154°C Flammepunkt: 82°C |
MDL Number:
MFCD00042356 CAS-nummer: 38565-52-5 EINECS: 254-004-4 |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...