2-(p-Tolyl)indole ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-(4-Methylphenyl)indole, 2-(4-Methylphenyl)-1H-indole

T3351-1G T3351-5G
TCIAT3351-1GEA 788 DKK
TCIAT3351-1G TCIAT3351-5G
2-(p-Tolyl)indole ≥98.0% (by GC)
2-(p-Tolyl)indole
Formel: C₁₅H₁₃N
MDL Number: MFCD00087262
CAS-nummer: 55577-25-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8b2cc16415d20c0e99a5a6453b8a947ac40548a&vid=d8b2cc16415d20c0e99a5a6453b8a947ac40548a&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 220.0 to 224.0 deg-C

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR