4-Tetrahydrothiopyranone-1,1-dioxide ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Tetrahydrothiopyran-4-one 1,1-Dioxide, 4-Tetrahydrothiopyranone 1,1-dioxide, 4-Oxothiane 1,1-Dioxide
T3320-200MG
T3320-1G
TCIAT3320-200MGEA
362
DKK
TCIAT3320-200MG
TCIAT3320-1G
4-Tetrahydrothiopyranone-1,1-dioxide ≥98.0% (by GC)
4-Tetrahydrothiopyranone-1,1-dioxide
Formel:
C₅H₈O₃S |
MDL Number:
MFCD00460698 CAS-nummer: 17396-35-9 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 170.0 to 174.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...