Tris acetate ≥99.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Tris(hydroxymethyl)aminomethane acetate salt, 2-Amino-2-(hydroxymethyl)propane-1,3-diol acetate salt, Trimethylolaminomethane acetate, Tris acetate, Tris(hydroxymethyl)aminomethane acetate, 2-Amino-2-(hydroxymethyl)-1,3-propanediol acetate

T3294-25G T3294-100G
TCIAT3294-25GEA 181 DKK
TCIAT3294-25G TCIAT3294-100G
Tris acetate ≥99.0% (by titrimetric analysis)
Tris acetate
Formel: NH₂C(CH₂OH)₃·CH₃COOH
Molekylvægt: 181.1870 g/mol
MDL Number: MFCD00038951
CAS-nummer: 6850-28-8
EINECS: 229-939-6

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 99.0 %
Purity (Neutralization titration) min. 99.0 %
Melting point 118.0 to 122.0 deg-C
Absorbance max. 0.500 (290 nm, 50%H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR